Lingettes pour salles blanches, Anticon® 100 StandardWeight™
Fournisseur: CONTEC
495352-803 495352-804 495352-805
115-0442EA 71.8 EUR InStock
115-0442 115-0443 115-3181
Lingettes pour salles blanches, Anticon® 100 StandardWeight™
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
100% no-run interlock knit polyester wipes with knife cut edges.
  • Patented textile technology creates a polyester wipe with high sorbency
  • Laundered and packed in a Class 4 cleanroom
  • Lowest fibres and particles available for a cut edge wipe to reduce risk of contamination
  • High sorptive capacity decreases the number of wipes required
  • Excellent choice for wiping and absorbing spills in critical cleanroom environments
Anticon 100® StandardWeight™ cleanroom wipes are made from 100% continuous filament polyester with a double-knit, no-run interlock construction. The clean cut edges create a very soft wipe, where the whole of the wipe fabric can be used with no risk of scratching. The wipes exhibit very low particle and fibre counts for a knife cut wipe, helping to minimise contamination. Ideal for spill control as well as applying disinfectants and cleaning solutions.
Recommended for ISO Class 3 - 5 environments.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité