Dispenser bucket, Wiper Bowl® Safe & Clean
Fournisseur: Zellstoff-Vertriebs-GmbH & Co. KG
60997-00
115-2744EA 10.4 EUR InStock
115-2744
Dispenser bucket, Wiper Bowl® Safe & Clean
Distributeurs de lingettes
This dispenser bucket made of plastic can be filled with wipe rolls and disinfectant or cleaning solutions. It is ideal for thorough cleaning applications in sensitive areas, e.g. in workshops. If a work area is disinfected simply close the lid and the remaining cloths will remain moist until their next use - for 30 days.
  • Refillabl with off-the-shelf disinfectants or cleaning agents
  • Economic and hygienic
  • Easy dispensing thanks to tear-off perforation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité