Dissipative floor finish, 10 L box, Statguard®
Fournisseur: DESCO
220521
115-3038EA 155 EUR InStock
115-3038
Dissipative floor finish, 10 L box, Statguard®
Produits nettoyants Détergents et Nettoyants de Surfaces
Statguard® Static Dissipative Cross-Linked Floor Finish is used to dissipate static charges as well as prevent triboelectric (static charge generation) charges while providing a clear, high gloss finish that resists wear. It meets EN 61340-5-1 ESD Floors Table 1 RG less than or equal to 1x10E9 ohms.
  • Non-tribocharging
  • Static dissipative
  • Converts a non-ESD floor to an ESD floor
  • Ensures worker safety and mitigates user’s liability exposure
  • Easy to apply; improves productivity
  • Cross-linked polymer; produces high-gloss, durable finish
  • Low humidity formulation
  • Made in the United States of America
Statguard® Static Dissipative Floor Finish is a free flowing liquid emulsion, which can be applied on any hard surface or sealed floor including vinyl, vinyl asbestos, linoleum, rubber, asphalt, sealed or painted wood, terrazzo and concrete. Statguard® technology eliminates the need for static control tile or floor mats. Statguard® dries in one hour and is ideal for cleanroom and electronic manufacturing, assembly, and test areas. This floor finish is for use in environments with relative humidity of 30-65%.
Statguard® provides superior electrical properties along with a safe walking surface. Underwriters Laboratory has evaluated Statguard® and tested it to their slip resistance standards. To ensure employee safety and to mitigate user’s liability exposure, it is important to use floor finish that has been successfully tested for slip resistance, and is properly installed and maintained.
Certifications: Lead-free RoHS compliant.
Conditionnement: Bag-in-box packaging.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité