Dissipative document wallets
Fournisseur: DESCO
238800
115-3061EA 5 EUR InStock
115-3061
Dissipative document wallets
Classeurs
Dissipative Document Wallets are rigid dissipative, low charging antistatic document vinyl wallets with self-adhesive backing are designed for use within ESD Protected Areas (EPA). Unprotected documents could hold electrostatic charges that may cause damage to electrostatic sensitive devices (ESDS).
  • The products contain antistatic additives that bloom to the surface of the film
  • The pouches have thumb holes and no markings.
The pouches are made of clear vinyl film that is low-charging and dissipative, which means that electrostatic charges will be removed to ground when handled by a grounded operator or placed on a grounded working surface.
The pouches use an adhesive that will not leave a residue on most surfaces. Density is 0,99 to 1,02 g/ccm. Auto-ignition temperature is above 250 ºC.
Film properties:
Density: 1.3 g/ccm
Cold break temperature: –20 ºC
Heat deflection temperature: +70 ºC
Film thickness and tolerance: 200 μm and ±10%
Outgassing 180 °C, <10E-5 Torr, 24 h: 0,53% TML / 0,09% CVCM
Surface resistance @ 22 °C / 15% rH (IEC 61340-2-3): <1×10¹¹ ohms
Charge decay time @ 22 °C / 15% rH (EIA 541 app. F): 0,1 s
12 day oven aging 70 ºC: Surface resistance <1×10¹¹ ohms, Charge decay time <2 s
Certifications: They meet or exceed the requirements for surface resistance of EN 61340-5-1 and Packaging standard IEC 61340-5-3.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité