Dual-wire thermoplastic wrist strap
Fournisseur: DESCO
4720
115-3067EA 10.1 EUR InStock
115-3067
Dual-wire thermoplastic wrist strap
Dispositifs de protection antistatique Bracelets antistatiques
The Dual-Wire thermoplastic wrist strap consists of an insulative exterior with a conductive interior and a 3,4 mm dual conductor jack. The wrist strap is lightweight, low profile, and safely makes contact with human skin.
  • Even if one dual-wire wrist strap conductor is severed, operator has reliable path-to-ground with other wire
  • Zipper style latching mechanism easily adjusts to most wrist sizes
Made to be used in conjunction with all SCS Workstation Monitors.
Certifications: Meets requirements of EN 61340-5-1 and Wrist Strap standard IEC 61340-4-6.
Informations de commande: Designed for use with SCS Dual-Wire Coil Cords (sold separately).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité