Elastic adjustable wrist straps, Jewel®
Fournisseur: DESCO
60001
115-3068EA 11.2 EUR InStock
115-3068
Elastic adjustable wrist straps, Jewel®
Dispositifs de protection antistatique Bracelets antistatiques
High visibility bright colour allows supervisor to see that worker is wearing wrist strap.
  • Adjustable
  • Locking clasp, band locked so it will not slip
  • Extra wide, soft elastic band for increased wearer comfort
  • Non-shedding, non-sloughing band material, safe for microelectronics
  • Nickel-plated brass wristband stud machined to exacting tolerances and precise profile
  • Silver impregnated polypropylene knitted on inside for superior conductivity
Replacement elastic bands available; and easy to replace without tools. Unique adjustment design eliminates exposed conductive 'tail' of band and provides added safety eliminating the chance of providing inappropriate ground path.
Medical grade 300 series stainless steel used for all metal parts which eliminates allergic skin reactions for most people.
Certifications: Coil cords are UL listed.
Informations de commande: Manufacturer's lifetime warranty on wrist strap stud.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité