Washroom cleaner and descaler, SURE®
Fournisseur: Diversey
100891583
115-3070EA 45.1 EUR InStock
115-3070
Washroom cleaner and descaler, SURE®
Produits nettoyants Détergents et Nettoyants de Surfaces
Concentrated, highly effective product for daily and periodical cleaning of water resistant hard surfaces in washrooms.
  • Concentrated formulation gives excellent economy of use
  • Effective cleaning and descaling even in hard water areas
  • Suitable for use on intact chrome and stainless steel surfaces
  • Cleans without leaving residue
  • No fragrance or dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for product in use for user
SURE® washroom cleaner and descaler is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certifications: OECD 301B test.
EU-Ecolabel (EU: NL/020/045).
Detergent Regulation, EC 648/2004.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité