11566.22
11566.36
11566.A3
11566.A9
11566.22EA
23.6
EUR
InStock
11566.22
11566.36
11566.A3
11566.A9
Ammonium sulfate ≥99.0% ACS
Ammonium sulphate
Purity: 99.0% min
CAS No.: 7783-20-2
Molecular Formula: H8N2O4S
Formula Weight: 132.14
Physical Form: Granular
Density: 1.769
Appearance: White
Melting Point: ca 280° dec.
Solubility: Soluble in water. Insoluble in alcohol and acetone
Merck Reference: 14,555
MDL No.: MFCD00003391
In galvanizing steel, manufacturing of ammonia alum and sulfuric acid