Synonyms:
Silver hexafluoroarsenate(V)
11608.09
11608.09EA
589
EUR
InStock
11608.09
Hexafluoroarsénate d'argent ≥98%
Hexafluoroarsénate d'argent
Purity: 98+%
CAS No.: 12005-82-2
Molecular Formula: AgAsF6
Formula Weight: 296.78
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Odour: Odourless
UN No.: 1557
MDL No.: MFCD00014142