SPINeasy® Plasmid Purification, MiniPrep Kit, MP Biomedicals
Fournisseur: MP Biomedicals
116534050
116534050.EA 149 EUR InStock
116534050.
SPINeasy® Plasmid Purification, MiniPrep Kit, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
SPINeasy Plasmid Miniprep kit is a silica-membrane spin-column kit that enables up to 20 µg of plasmid DNA to be purified from 1 to 5 ml of transformed bacterial culture.
High purity plasmid DNA can be obtained within 25 minutes through a simple purification process and is immediately ready for routine molecular biology laboratory applications.
Key applications are In-vitro transcription, cloning, NGS, PCR, qPCR, restriction digestion, transformation, genomics, molecular biology.
Certifications: Hazard Class 8
Information de livraison: Kit includes 20 ml Resuspension Buffer SN1, 20 ml Alkaline Lysis Buffer SN2, 30 ml Neutralization Buffer SN3, 21 ml Wash Buffer NW1, 6 ml Wash Buffer NW2, 10 ml Elution Buffer SN, 200 µL RNase A, 50× Column SN with collection tubes.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité