SPINeasy® DNA Extraction Kit for Plant, MP Biomedicals
Fournisseur: MP Biomedicals
116535050
116535050.EA 434 EUR InStock
116535050.
SPINeasy® DNA Extraction Kit for Plant, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
SPINeasy DNA Kit for plant is a silica-membrane spin-column kit that enables quick, convenient and cost-effective isolation of genomic DNA (gDNA) from various plant tissues, such as leaf, stem and seed.
High quality gDNA can be extracted from polysaccharide and polyphenolic-rich plants. The use of our specially formulated Lysis Buffer PD and Lysing Matrix A in combination with FastPrep® Instruments from MP Biomedicals enables highly efficient lysis of various types of samples within seconds. For DNA purification, Column PD and kit buffers are designed to deliver gDNA extracts of high yield and purity which are compatible with downstream molecular biology applications such as PCR, restriction digestion and sequencing.
Key applications are plant genomics, molecular biology, PCR, NGS.
Certifications: Hazard Class 8
Information de livraison: Provided with 50×2 ml Lysing Matrix A, 50 ml Lysis Buffer PD, 12 ml PPS Solution, 15 ml Binding Buffer PD, 2×12 ml Wash Buffer, 10 ml Elution Buffer PD, 0,8 ml RNase A, 50×Column PD with collection tubes.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité