SPINeasy® PCR Purification and Gel Extraction Kit, MP Biomedicals
Fournisseur: MP Biomedicals
116538050
116538050.EA 133 EUR InStock
116538050.
SPINeasy® PCR Purification and Gel Extraction Kit, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
SPINeasy PCR purification and gel extraction Kit is a silica-membrane spin-column kit that enables quick and convenient DNA clean up from various enzymatic reactions, such as PCR and restriction digestion, as well as isolation and purification of DNA fragments from agarose gel electrophoresis.
Upto 23 µg of DNA of molecular weight ranging from 100 bp to 20 kb can be purified through a quick and simple process. Purified DNA is immediately ready for routine molecular biology laboratory applications.
SPINeasy PCR purification and gel extraction kit is a silica-membrane spin-column kit that enables quick and convenient DNA clean up from various enzymatic reactions, such as PCR and restriction digestion, as well as isolation and purification of DNA fragments from agarose gel electrophoresis.
Certifications: Hazard Class 8
Information de livraison: Kit includes 50 ml Binding Buffer PG, 6 ml Wash Buffer PG, 10 ml Elution Buffer PG, 50× Column PG with collection tubes.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité