SPINeasy® Plasmid Purification, MidiPrep Kit, MP Biomedicals
Fournisseur: MP Biomedicals
116539025
116539025.EA 385 EUR InStock
116539025.
SPINeasy® Plasmid Purification, MidiPrep Kit, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
SPINeasy Plasmid Midiprep kit is a silica-membrane spin-column kit for the purification of high-copy and low-copy number plasmid DNA from 25 to 50 ml of transformed bacterial (E.coli) culture.
Using our specially formulated buffers, bacterial cells are disrupted by alkaline lysis to release plasmid DNA. Cell debris, such as chromosomal DNA and proteins, are precipitated by neutralization and cleared through centrifugation. Subsequently, plasmid DNA in the lysate supernatant is adsorbed on a silica membrane, washed and eluted in a convenient process using the spin column technology. Upto 1 mg of plasmid DNA can be purified through this simple workflow, which is immediately ready for routine molecular biology laboratory applications.
Key applications are In-vitro transcription, cloning, NGS, PCR, qPCR, restriction digestion, transformation, genomics, molecular biology.
Certifications: Hazard Class 8
Information de livraison: Kit includes 60 ml Resuspension Buffer SD1, 60 ml Alkaline Lysis Buffer SD2, 90 ml Neutralization Buffer SD3, 70 ml Wash Buffer DW1, 12 ml Wash Buffer DW2, 30 ml Elution Buffer SD, 600 µl RNase A, 25× Column SD with collection tubes.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité