Lysing Matrix B, MP Biomedicals
Fournisseur: MP Biomedicals
116540425
116540425.EA 270 EUR InStock
116540425.
Lysing Matrix B, MP Biomedicals
Homogénéisateurs Homogénéisateurs de paillasse
Lysing Matrix B tubes are used primarily for lysis of gram positive and gram negative bacteria, and will also disrupt fungal tissue and spores
  • Impact resistant tube containing 0.1mm silica spheres
  • Can be used as an additive for disruption by sonication
  • Use for gram-positive and gram-negative bacteria
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under a rigorous set of proprietary conditions that offer customers a premium product that can be immediately used with confidence.Matrices are available separately for use with your own unique buffers, and are also available as components of the complete purification kits.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité