SPINeasy RNA kit for bacteria, 50 preparations, MP Biomedicals
Fournisseur: MP Biomedicals
116541050
116541050.EA 456 EUR InStock
116541050.
SPINeasy RNA kit for bacteria, 50 preparations, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
Quick and convenient purification of total RNA from gram-positive and gram-negative bacteria using spin column technology.
  • The kit allows multiple samples to be processed Simultaneously and without the use of toxic substances such as phenol and chloroform
  • Total RNA of high quality and integrity can be purified typically within 40 minutes is immediately available for RT-PCR and other downstream applications
SPINeasy RNA Kit for Bacteria is a silica-membrane spin-column kit that enables quick and convenient purification of total RNA from gram-positive and gram-negative bacteria. Included in the kit is our specially formulated RNASS solution that stabilizes and protects RNA in bacteria samples.
The use of Lysis Buffer R and Lysing Matrix B in combination with FastPrep® instruments from MP Biomedicals enables highly efficient lysis of bacterial samples within seconds.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité