SPINeasy® DNA Pro Kit for Feces, MP Biomedicals
Fournisseur: MP Biomedicals
116547050
116547050.EA 448 EUR InStock
116547050.
SPINeasy® DNA Pro Kit for Feces, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
The SPINeasy® DNA Pro Kit for Feces is able to handle a wide range of fecal samples.
  • Newly formulated buffers to achieve better yield and purity of fecal DNA
  • Easy to handle and minimise any risk of contamination
  • Suitable for various types of feces samples
  • User friendly, suitable for any scale of experimental throughput
The DNA yield, purity (260/280 and 260/230 ratio) and integrity were assessed using spectrophotometer and DNA gel. Each dot of the plot represents a single extraction. The horizontal bars indicate the median value.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité