SPINeasy® DNA Purification Kit, MP Biomedicals
Fournisseur: MP Biomedicals
116548050
116548050.EA 198 EUR InStock
116548050.
SPINeasy® DNA Purification Kit, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
Humic acids, heavy metals, heme are the most notorious PCR inhibitors. This could be due to non-optimised DNA purification procedures which often co-purify inhibitors and lead to false negative results on downstream applications.
  • Efficient clean-up of highly contaminated pre-purified DNA sample
  • Clean: Effective removal of inhibitory compound
  • Buffer P1 and P1HA suitable for sample with various degree of contamination
The SPINeasy® DNA Purification Kit is formulated to effectively remove contaminants on pre-purified DNA samples using the novel and proprietary humic acid removal technology. It can recover high quality DNA for all downstream applications.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité