SPINeasy® DNA Kit for Blood, MP Biomedicals
Fournisseur: MP Biomedicals
116552050
116552050.EA 179 EUR InStock
116552050.
SPINeasy® DNA Kit for Blood, MP Biomedicals
Réactifs pour acides nucléiques Nucleic Acid Purification Kits and Reagents
SPINeasy DNA kit for blood is a high-performance genomic DNA (gDNA) extraction kit based on silica-membrane spin-column technology. This kit enables quick isolation of gDNA from whole blood preserved in different anticoagulants (EDTA, Heparin and sodium citrate), typically in less than 30 minutes.
  • Rapid and efficient DNA isolation in less than 30 minutes
  • Fresh, frozen, or anticoagulated blood
  • Simple and effective lysis protocol using Lysis buffer BL and Proteinase K to omit the need of any mechanical lysis
  • No organic extraction
  • High purity DNA ready to use for downstream application
It also allows easy gDNA isolation from other samples, including plasma, serum, saliva, and cell culture medium. The use of our specially formulated Lysis buffer BL omits the need of homogenization and enables highly efficient lysis of various samples. Provided in the kit, Column S and kit buffers are designed to deliver gDNA extracts of high yield and purity, compatible with downstream applications such as PCR amplification.
Key applications are Genomics; Molecular biology; PCR, qPCR, Microbiome analysis, NGS; shotgun sequencing.
Sample colume 200 μl of Human blood, serum, plasma, saliva, or cell culture media.
Information de livraison: Kit includes Lysis buffer BL, Wash buffer BW1, Wash buffer BW2, Elution buffer EB, Column S, 2,0 ml collection tubes, 1,5 ml microcentrifuge tubes, Proteinase K, Proteinase K buffer.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité