Packaged under argon.
12381.FI
12381.FIEA
477
EUR
InStock
12381.FI
Dysprosium ≥99.9% (REO, base d'oxyde de terre rare), en feuilles, Thickness: 0.25 mm (0.01 in)
Dysprosium
Purity: 99.9%
CAS No.: 7429-91-6
Physical Form: ≈1.33g/25x25mm, Packaged under argon
Odour: Odourless
UN No.: 3178
MDL No.: MFCD00010982