Packaged under argon.
12388.FF
12388.FFEA
404
EUR
InStock
12388.FF
Erbium ≥99.9% (REO, base d'oxyde de terre rare), en feuilles, Thickness: 0.025 mm (0.001 in)
Erbium
Purity: 99.9%
CAS No.: 7440-52-0
Physical Form: ≈0.14g/25x25mm, Packaged under argon
UN No.: 3178
MDL No.: MFCD00010987