Estradiol Cypionate System Suitability Mixture, Reference Standard
Fournisseur: USP
1252193
1252193.EA 1230 EUR InStock
1252193.
Estradiol Cypionate System Suitability Mixture, Reference Standard
Étalons Étalons organiques
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
This mixture contains estradiol cypionate, estradiol-9-ene cypionate, estradiol dicypionate, and 4-methylestradiol cypionate
Certifications: US Pharmacopea Reference Standard
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité