Sweeper
Fournisseur: FHP VILEDA SCS
119908
129-0459EA 29.9 EUR InStock
129-0459
Sweeper
Balais
The Sweeper and Pan is the perfect system for ergonomic and efficient dirt and liquid removal.
  • Flexible joint allows the user to reach into the most narrow spaces
  • Sweep up broken glass in your cleanroom quickly and easily
  • No need to change gloves when an item is dropped on the floor
  • Keeps workers from touching the floor within the cleanroom
  • Can be sprayed or wiped down with common disinfectants
  • Stands up easily for storage or can be hung
  • Adjustable bag holder for different size collection bags
The system is hygienic because of the washable and non-corroding material used in the construction. Overlapping rubber at the edges of the squeegee protect walls and furniture and the ergonomic grip reduces worker fatigue. Perfect for dry or damp conditions on smooth floors.
Informations de commande: Sweeper and pan have to be ordered separately.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité