Sharps containers, code YELLOW, SHARPAK™
Fournisseur: Synergy Health
SU02230 SU02430 SU02630 SU03530
129-1052EA 10.1 EUR InStock
129-1052 129-1055 129-1058 129-1060
Sharps containers, code YELLOW, SHARPAK™
Poubelles Collecteurs de déchets tranchants
All SHARPAK™ Code YELLOW sharps containers feature yellow coloured closures and are recommended for the segregation, collection and disposal of partially discharged sharps waste which are NOT contaminated with cytotoxic or cytostatic medicines.
  • Yellow colour coded closures to meet the recommendations on waste segregation as detailed in the Department of Health's technical memorandum HTM-07-01
  • Secure closure with locking facility
  • Distinctive colouring and clear labelling
  • Sequential numbering for traceability
  • Supplied assembled
Code YELLOW SHARPAK™More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité