Sterilisation flat pouches, Stericlin®
Fournisseur: VP Medical Packaging
3FKFB250110 3FKFB250105 3FKFB250115 3FKFB250120 3FKFB250125
129-1297EA 448 EUR InStock
129-1297 129-1296 129-1298 129-1299 129-1304
Sterilisation flat pouches, Stericlin®
Sachets Poches de stérilisation
Stericlin® sterilisation pouches, Tyvek® 1073B with see-through film and indicators for VH₂O₂ (plasma) and EO.
  • Nonwoven polyethylene
Barrier properties against germs and moisture, extreme high puncture resistance and fiber-free peel without delamination draw this sterile barrier system. Suitable for steam sterilisation, but absolutely compatible with the VH₂O₂ (plasma) and radiation sterilisation.
Tyvek® and the composite film used are ideal for VH₂O₂ (plasma), EO, FORM and irradiation sterilisation. Two indicators for the VH₂O₂ (plasma) and EO sterilisation are printed outside the filling area.
The stericlin® sterile barrier system is first choice in gas sterilisation, as Tyvek® and the composite film hardly absorb the toxic gases and enable residual gases to be far below specified limits.
Certifications: DIN EN ISO 11607 and DIN 868-5.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité