Sterilisation flat reels, Tyvek®/film, Stericlin®
Fournisseur: VP Medical Packaging
3FKFS250206 3FKFS250212 3FKFS250214 3FKFS250216 3FKFS250218 3FKFS250222
129-1306EA 443 EUR InStock
129-1306 129-1308 129-1309 129-1310 129-1311 129-1312
Sterilisation flat reels, Tyvek®/film, Stericlin®
Sachets Poches de stérilisation
These see-through sterilisation reels are made of a medical grade of Tyvek® non woven polyethylene fabric, Type 1073 B with 75 g/m².
  • Effective barrier against germs, bacteria and moisture
  • High resistance to penetration and fibre-free peeling
  • Not suitable for steam sterilisation
This is fully compatible with the requirements of VH₂O₂ (plasma) sterilisation and irradiation. This sterile barrier system is the first choice for gas sterilisation, for Tyvek® hardly absorbs any toxic gases and thus results in residual toxic gas levels that are far below the legal limit values.
Certifications: ISO 11607. CE.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité