BRUTE® Self-Draining, Dome Top, and Funnel Top Lids
Fournisseur: Rubbermaid Commercial Products
FG263100RED FG263100YEL FG264560GRAY FG354300GRAY FG264788RED FG263788RED FG264560BLA FG261960BLA FG264560WHT FG263100GRAY FG265400GRAY FG263100WHT FG263100BLUE FG260900GRAY FG260900WHT FG261960GRAY FG261960WHT 1779731 FG264560RED
129-1852EA 29.1 EUR InStock
129-1852 129-1850 129-1882 129-1842 129-1878 129-1889 129-1883 129-1866 129-1880 129-1853 129-1877 129-1851 129-1855 129-1870 129-1869 129-1864 129-1863 129-2172 129-1881
BRUTE® Self-Draining, Dome Top, and Funnel Top Lids
Poubelles Conteneurs de déchets
Select from a wide range of lids for use in any environment.
  • Choose from self-draining, dome top, or funnel top lid styles
  • Snap-lock closures ensure a secure fit on compatible containers
Self-Draining lids feature self-draining channels to prevent water from pooling, and to reduce still water build-up. A snap-lock feature ensures a secure fit, and ridges enable stacking of BRUTE® containers to provide a space-efficient storage solution. These lids are available in 7 colours.
Dome Top lids feature a spring-action door to allow easy trash disposalMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité