Humidity indicator 3 spot cards
Fournisseur: DESCO
51060HIC125
129-2643EA 22.5 EUR InStock
129-2643
Humidity indicator 3 spot cards
Papiers test Papiers indicateurs
Humidity indicator cards (HICs) are printed with moisture sensitive spots which respond to various levels of humidity with a visible colour change from blue to pink.
  • The moisture sensitive spots are made of blotting paper impregnated with cobaltous chloride
  • The humidity inside dry packed barrier bags can be monitored by including an HIC inside
When the bag is opened, the card can be examined for proper dryness inside the bag.
L×W: 76,2×50,8 mm (3×2 ").
Certifications: Complies with IPC/ JEDEC J-STD-033D.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité