Synonyms:
Tetracobalt dodecacarbonyl
13055.06
13055.06EA
438
EUR
InStock
13055.06
Dodecacarbonyltetracobalt ≥98%
Dodecacarbonyltetracobalt
Purity: 98% min
CAS No.: 17786-31-1
Molecular Formula: C12Co4O12
Formula Weight: 571.86
Physical Form: Crystalline
Density: 2.09
Appearance: Black
Melting Point: 60° dec.
UN No.: 3179
MDL No.: MFCD00064698