Armoires de laboratoire, sans étincelle, série EX
Fournisseur: Lovibond
2422105-V
132-0950EA 1300 EUR InStock
132-0950
Armoires de laboratoire, sans étincelle, série EX
Mobilier Armoires de stockage
The cabinet consist of a sturdy sheet steel housing with impact-proof and jolt-resistant powder coating. Improved, robust, insulated housing and highly efficient components provide maximum energy efficiency. The robust interior is made of high-quality, strong white plastic material (PS). The door is lockable and supplied with a right-hand hinge as standard (but can easily be converted to a left-hand hinge). A tight door seal is ensured by an all-round magnetic gasket.
  • Lockable door, changeable door stop
  • Temperature regulation: continuous 1 to +15 °C
  • Cooling: powerful compressor unit, mounted on low noise, vibration-free bearings. Coolant: R600a
  • Automatic defrost - condensation drains into a collection bowl within the refrigerator
  • Adjustable front feet
The models EX 220, EX 300 and EX 490 have a 'fan stop' function, which switches the fan off when the door is opened.
Certifications: The product complies with the following european directives and regulations: 2006/42/EC, 2006/95/EC, 94/9/EC, 2004/108/EC, 2011/65/EU.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité