13444.36
13444.36EA
93.8
EUR
InStock
13444.36
Ammonium dichromate ≥99.5% ACS
Ammonium dichromate
Purity: 99.5% min
CAS No.: 7789-09-5
Molecular Formula: (NH4)2Cr2O7
Formula Weight: 252.10
Physical Form: Crystalline
Odour: Odourless
Merck Reference: 14,515
UN No.: 1439
MDL No.: MFCD00010879
Catalysts, source of pure nitrogen, in lithography, manufacturing of pigments