Safety cabinets with storage shelves (CLASSIC line standard) or pull-out shelves (CLASSIC line pro) for the storage of hazardous substances in working spaces. Complete carcass, interior fittings and wing doors are made of powder-coated sheet steel.
- Doors with standard integrated arrest system, meaning that containers can be placed into and removed from the cabinet with both hands
- Removable base and adjustable feet for easy alignment and transport with a hand pallet truck.
- The base is set back to prevent hitting one’s feet
- Ventilation at every cabinet level, ability to visually check the venting cut-off flaps ; the cut-off flaps close automatically in case of fire
Safety cabinets are fire resistant up to 90 minutes. In case of fire, a thermocouple initiates the closing of the doors.
Standard equipotential bonding saddle on the roof of the cabinet for connection to the earthing in accordance with TRGS 727 (avoiding ignition hazards). The interior fittings are conductively connected with the equipotential bonding saddle.
The diameter of the exhaust air connection socket is 75 mm.
Certifications: Type-tested by MPA/ TÜV SÜD according to DIN EN 14470 1, laboratory furniture standard DIN EN 14727, EK5/AK4 09-10:2009 and TRGS 510 Annex 3.
Classification Type 90 in accordance with DIN EN 14470-1, GS/CE mark and High Quality mark of approval by TÜV SÜD for upper level style, user-friendliness and increased lifetime.
Information de livraison: Delivery is not included, prices available on request.