Ductless fume hoods, Paramount®
Fournisseur: LABCONCO
6963320
135-2023EA 0 EUR InStock
135-2023
Ductless fume hoods, Paramount®
Hottes et coffrets Hottes filtrantes
Self-contained ductless enclosures allow safe handling of organic, formaldehyde, acid gas, sulphur compounds, or ammonia chemical fumes and vapours, as well as particulates. Air inside the enclosures passes through carbon filters, and in some cases additional HEPA particulate filters, before being safely exhausted back into the laboratory. No outside ducting is required.
  • Telescoping base stands adjust in seven height positions from 69,9 to 75,1 cm
  • Glacier white stands are constructed of 4,4 cm tubular epoxy-coated steel
  • Hydraulic lift base stands are
More Product Information
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité