Synonyms:
Lanthanum trifluoride
13663.18
13663.18EA
186
EUR
InStock
13663.18
Trifluorure de lanthane, anhydre ≥99.9% (REO, base d'oxyde de terre rare), REacton®
Trifluorure de lanthane
Purity: 99.9%
CAS No.: 13709-38-1
Molecular Formula: F3La
Formula Weight: 195.90
Physical Form: Lump
Density: 5.936
Melting Point: 1493°
MDL No.: MFCD00011073