Lonicera Macranthoides Flower Dry Extract, Reference Standard
Fournisseur: USP
1369703
1369703.EA 159 EUR InStock
1369703.
Lonicera Macranthoides Flower Dry Extract, Reference Standard
Étalons Étalons organiques
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Mixture, with identified compendial components Dipsacoside B, Macranthoidin A and Macranthoidin B
Certifications: US Pharmacopea Reference Standard
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité