Miller® DuraFlex® AGU 90S Elasticated Harness
Fournisseur: HONEYWELL SAFETY
ME13344
139-0243EA 230 EUR InStock
139-0243
Miller® DuraFlex® AGU 90S Elasticated Harness
Protections anti-chutes Harnais de sécurité
Four point safety harness with integrated positioning belt, rear fall arrest eyelet and 2 retaining eyelets.
  • Made from DuraFlex® (highly elastic stretch fabric): adapts to the user's body shape and expands with every movement so that the straps do not dig in, hinder or get twisted
  • Additionally treated with PTFE
  • Additional arrester eyelet, all eyelets made from stainless steel
  • Quick-release closures
  • Unrivalled comfort and freedom of movement as well as a soft catch in the event of a fall
Certifications: EN 361.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité