Hupfer Variocart® MVC 8×6/3 KST/gr Transport Trolley
Fournisseur: HUPFER METALLWERKE
MVC8X6/3KST/GR
139-0818EA 846 EUR InStock
139-0818
Hupfer Variocart® MVC 8×6/3 KST/gr Transport Trolley
Chariots
Demountable transport trolley made of high-quality stainless steel with deep shelves.
  • Height grid: 66,5 mm
  • On-board loading: 100 kg
Demountable transport trolley in a sturdy, self-supporting and hygienic design made of high-quality stainless steel round tube frame Ø25 mm with adjustable, height-adjustable shelves, bevelled on all sides, with a sturdy clamping device. Four polyethylene disc bumpers serve as collision protection. Runs on four corrosion-resistant swivel castors Ø125 mm, two with locks, grey polymer housing with air-identic polymer wheel and corrosion-resistant ball bearing.
The shelves can be individually expanded and replaced. The shelves can be individually expanded and replaced. The Variocart is versatile, but its dimensions make it particularly ideal for Euro containers.
Due to the flatpack carton, the Variocart requires 75% less storage and transport space.
Furthermore, dirt on the castors is avoided (thus delivery of unused rolls).
Information de livraison: The Hupfer Variocart is packed in a flatpack carton and can therefore be sent with a parcel service. Furthermore, a total of 12 Variocarts can be shipped per Euro Pallet.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité