Aspirateur pour salles blanches, MV-1CR (CG)
Fournisseur: TIGER-VAC
110739B
141-0955EA 2540 EUR InStock
141-0955
Aspirateur pour salles blanches, MV-1CR (CG)
Aspirateurs
Designed for general clean up in contamination controlled environments. Cleanroom compatible, can be used in the pharmaceutical industry.
Contruction: Stainless steel AISI 304.
  • Four-stage filtration system includes a ULPA filter (U15); double cloth filter, dome filter and disposable filter bag
  • Static-free
  • Special filter lined with stainless steel is available for the recovery of broken wafers and sharp objects
  • Single phase through-flow motor
  • Recovery capacity of 5,7 litres in disposable filter bag
  • Equipped with a golf cart for easy maneuverability
This unit is equipped with a true ULPA Filter (U15) located upstream of the motor. It is EMI/RFI shielded, equipped with an electronic filter suppresser which reduces electromagnetic interference (EMI), it provides shielding against RFI (Radio Frequency Interference), and will not disrupt computers and other sensitive electronic equipment. This unit is static-free and ESD safe. Any static produced while vacuuming is quickly and effectively discharged to the ground.
Note: For dry recovery only.
ULPA vacuum cleaner is aerosol leak tested.
Cleanroom compatible with ISO Class 4 (former Class 10).
Certifications: ULPA vacuum cleaner is aerosol leak tested.
Cleanroom compatible with ISO Class 4 (former Class 10).
Information de livraison: Complete tool kit included.
Attention: Note: For dry recovery only.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité