Balais trapèze, AlphaMop™, PE
Fournisseur: Texwipe
TX7108 TX7118 TX7026 STX7118 TX7118M TX7118F TX7108AH TX7108H TX7123 STX7118M
129-0180EA 148 EUR InStock
129-0180 129-0181 141-0249 141-1048 141-1060 141-1059 141-1056 141-1057 141-1064 141-1049
Balais trapèze, AlphaMop™, PE
Serpillières
Ideal for cleaning ceilings, walls, floors, and other flat surfaces in critical environments. The changeable, laundered polyester head fits over a flat foam swivel pad assembly attached to a fibreglass handle. Replaceable foam pad ensures the polyester head conforms to textured floors and other surfaces while resisting corrosion and abrasion. A lightweight, durable fibreglass handle gives the user excellent control over the swivel head.
  • Lightweight mop maintains ultra-low generation of particles and extractables
  • Mop can be sterilised
  • Mophead covers can be pre-wetted with 6% isopropanol
  • Accesses hard-to-reach areas with ease
  • Excellent durability with low contamination
  • Allows user to change covers and foam pads frequently, adhering to wiping protocol
  • Rapid-change cover and foam pad configuration
* Handle length.
Conditionnement: En salle propre, sachets doubles selon le système Bag-Within-A-Bag®.
Attention: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité