Glass cleaners, SURE®
Fournisseur: Diversey
100891496
141-1570EA 31.9 EUR InStock
141-1570
Glass cleaners, SURE®
Produits nettoyants Détergents et Nettoyants de Surfaces
Ready to use spray cleaner for streak-free glass cleaning.
  • Excellent removal of fingermarks as well as the spot treatment of other slightly adherent soils
  • Easy spray-on, wipe-off action
  • No fragrance nor dye
  • Contains plant-based ingredients from renewable sources
  • Decomposed by natural processes
  • No environmental hazard classifications, no safety warnings for product for user
SURE® glass cleaner is part of a unique range of plant-based, Cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certifications: OECD 301B test
EU-Ecolabel (EU: NL/020/045)
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité