Synonyms:
Barium i-propoxide
14523.14
14523.14EA
555
EUR
InStock
14523.14
Barium isopropoxide
Barium isopropoxide
CAS No.: 24363-37-9
Molecular Formula: C6H14BaO2
Formula Weight: 255.52
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Brown to yellow
Melting Point: ca 200° dec.
UN No.: 3205
MDL No.: MFCD00050486