Synonyms:
Holmium(III) fluoride, Lutetium (III) fluoride
14620.06
14620.06EA
300
EUR
InStock
14620.06
Trifluorure de lutetium ≥99.9% (REO, base d'oxyde de terre rare), REacton®
Trifluorure de lutetium
Purity: 99.9%
CAS No.: 13760-81-1
Molecular Formula: LuF3
Formula Weight: 231.97
Physical Form: Lump
Density: 8.3
Melting Point: 1182°
UN No.: 3288
MDL No.: MFCD00016191