Detergent for use in immersion baths, liquid concentrate, neodisher® LM 10
Fournisseur: Dr. Weigert
440130
148-3177EA 281 EUR InStock
148-3177
Detergent for use in immersion baths, liquid concentrate, neodisher® LM 10
Produits nettoyants Détergents et Nettoyants de Surfaces
Manual cleaning of laboratory glassware in immersion baths in medical and biological research, laboratories in the food industry. Manual cleaning of cages for experimental animals by immersion.
  • Viscosity: <10 mPas (concentrate, 20 °C)
  • Density: 1,2 g/cm³ (20 °C)
  • Titration factor: 1,90
  • Sensitive to frost below –5 °C
neodisher® LM 10 is suitable for precleaning items to be washed which will then undergo a main mechanical cleaning process. It removes radioactive contaminants and organic residues, which are very stubborn and also suitable for pre-cleaning of cages for experimental animals. All usual glass, porcelain and ceramic materials are safe from attack by neodisher® LM 10 solutions. If rubber, plastic and metallic instruments are being washed, their resistance must be checked. It can be used with all degrees of water hardness.
Contains <5% chlorine-based bleaching agents, 5 to 10% phosphonates according to regulation (EC) No 648/2004 on detergents.
Attention: Do not mix with other products. Only for professional use. This product is sensitive to heat and sunlight. Store in a cool place.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité