Laboratory glass cleaner, deconex® 21 SOLID
Fournisseur: Borer Chemie
501100.00-S10P
148-5191EA 106 EUR InStock
148-5191
Laboratory glass cleaner, deconex® 21 SOLID
Produits nettoyants Détergents et Nettoyants de Surfaces
Mildly alkaline cleaning powder for automated cleaning of laboratory glassware und reusable laboratory material.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • With a broad cleaning spectrum and excellent results even at low concentrations
  • High cleaning performance due to excellent dispersing capabilities
  • Good material compatibility, especially recommended for alkali and corrosion sensitive items, also suitable for aluminium
  • EDTA-, NTA-, chlorine- and surfactant-free, no release of surfactants into waste water
  • Rinsing with an acidic deconex® cleaner is recommended as the completion of the cleaning process
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité