VWR®, Lab Drying Rack
Fournisseur: VWR Collection
149-0297EA 268 EUR InStock
149-0297
VWR®, Lab Drying Rack
Portoirs Égouttoirs
Versatile rack made of durable, easy to clean PS.
  • Channels designed for more useful configurations
  • Use on counter or mount on wall
  • Large and small pegs for all glassware varieties
  • One of a kind drying basket to collect small items
Designed to protect against breakage
- Peg angle allows for better hold - especially for large, heavy pieces
- Patent pending SureGrip hook securely grips glassware into place
- Holds up to 35 kg
Information de livraison: Includes 72 pegs (36 small, green and 36 large, blue), 2 drying baskets (clear), drainage tubing,More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité