Gas detection tubes for nitrogen dioxide
Fournisseur: Dräger
6719101
151-0140EA 108 EUR InStock
151-0140
Gas detection tubes for nitrogen dioxide
Tubes de détection de gaz
To measure the 'spot concentrations' of nitrogen dioxide for use with an Accuro® pump. Uses include determination and measurement of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.
  • Within their TLV, ozone or chlorine do not interfere with the reading
  • Higher concentrations indicated however with differing sensitivity period
  • Nitrogen monoxide is not indicated
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuring range is calibrated at 20 °C and 1013 hPa.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité