Cell culture reagents, microcarriers, Cytodex 3
Fournisseur: Cytiva
17-0485-01 17-0485-03 17-0485-02
17-0485-01EA 377 EUR InStock
17-0485-01 17-0485-03 17-0485-02
Cell culture reagents, microcarriers, Cytodex 3
Perles Billes microporteuses
Cytodex 3, formed by chemically coupling a thin layer of denatured collagen to the cross-linked dextran matrix, is the microcarrier of choice for cells that may be difficult to culture in vitro, culture of primary cells or those with an epithelioid morphology, especially cells with low plating-efficiency or differentiated or sensitive cell-types which are normally difficult to grow, e.g. hepatocytes, endocrine cells.
  • Contains a layer of acid-denatured porcine collagen (gelatin) on the surface of the microporous matrix
  • Designed to yield harvested cells of
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité