Reversed phase chromatography media, SOURCE™ 15RPC
Fournisseur: Cytiva
17-0727-20
17-0727-20EA 423 EUR InStock
17-0727-20
Reversed phase chromatography media, SOURCE™ 15RPC
Protéines purification Protein Purification Resins
SOURCE™ 15RPC is designed for reversed phase chromatography (RPC) with full pH range stability and suitable for polishing steps in industrial processes requiring high flow velocities and low back-pressure.
  • Mono-sized 15 µm rigid beads gives low back pressures and high flow rates allowing for high productivity
  • Designed for high resolution reversed phase chromatography of proteins, peptides and oligonucleotides
  • High chemical stability allowing for wide range of working conditions with good resistance to cleaning conditions at high pH
  • Bioprocess medium supported for industrial applications
A controlled pore size distribution and large specific surface area offer excellent capacity for a wide range of molecules, from small peptides and oligonucleotides up to large proteins. With full pH range stability, SOURCE™ 15RPC is a strong alternative to silica-based RPC matrices.
Available in a range of different bulk pack sizes for easy scale-up and process development.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité