Gelatin-Sepharose® 4B
Fournisseur: Cytiva
17-0956-01
17-0956-01EA 372 EUR InStock
17-0956-01
Gelatin-Sepharose® 4B
Protéines purification Protein Purification Resins
Gelatin Sepharose® 4B affinity media with immobilised gelatin is designed for the purification or depletion of fibronectin as gelatin binds to fibronectin with high specificity. Gelatin Sepharose® 4B is bovine gelatin coupled to Sepharose® 4B by the cyanogen bromide method.
Gelatin binds specifically to fibronectin, which is a high molecular weight glycoprotein found on the surfaces of many cell types and present in many extracellular fluids including plasma. Gelatin Sepharose 4B has been designed for the purification or removal of fibronectin.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité