Hydrophobic interaction chromatography columns, HiTrap™ Butyl-S-FF Sepharose™
Fournisseur: Cytiva
17-0978-13
17-0978-13EA 238 EUR InStock
17-0978-13
Hydrophobic interaction chromatography columns, HiTrap™ Butyl-S-FF Sepharose™
Colonnes pour chromatographie
HiTrap™ Butyl-S-FF are prepacked Butyl-S Sepharose™ 6 Fast Flow columns for hydrophobic interaction chromatography (HIC).
  • Butyl-S Sepharose™ 6 Fast Flow is well-established and is the least hydrophobic medium in the Sepharose™ Fast Flow HIC range
  • Can be used with syringes, single laboratory pump or chromatography system such as ÄKTA design
  • High flow rates and high capacities
  • Excellent and easy scale-up achieved by connecting several HiTrap™ columns in series or using HiPrep™ columns
The columns are used for small-scale protein purification as well as for screening of binding and elution conditions.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité