Anion exchange chromatography columns, HiTrap Q HP
Fournisseur: Cytiva
17-1153-01 17-1154-01 29-0513-25
17-1153-01EA 176 EUR InStock
17-1153-01 17-1154-01 29-0513-25
Anion exchange chromatography columns, HiTrap Q HP
Colonnes pour chromatographie
HiTrap® Q HP are prepacked, ready-to-use High Performance strong anion exchange columns for high-resolution, small-scale protein purification.
  • Convenient and reproducible for fast, easy, high-performance separations either alone or connected in series
  • 34 µm bead size for high-performance, high-resolution purifications
  • Strong quaternary ammonium (Q) anion exchanger
  • Designed for use with a syringe, peristaltic pump, and chromatography systems such as ÄKTA design
The Q functional group is a quaternary amino group coupled to the matrix via chemically stable ether linkages. Q Sepharose High Performance is a strong anion exchanger charged with high loading capacity over broad pH ranges.
HiPrep Q HP anion exchange columns are used in a variety of applications, including IEX chromatography during research on subdomains of fragments of bacterial endoglycosidase S and immunoglobulin G.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité